COM456879050 Combi-Blocks, Inc.

2-Hydroxy-5-(methoxycarbonyl)benzoic acid

  • MDL number MFCD16987712
  • Cas Number : 79128-78-2
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5832
Availability details for
Quantity
  • Availability
  • Add to Cart