COM456879054 Combi-Blocks, Inc.

Methyl 4-[(dimethyl-1,2-oxazol-4-yl)methoxy]benzoate

  • MDL number MFCD02329782
  • Cas Number : 315693-84-6
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5835
Availability details for
Quantity
  • Availability
  • Add to Cart