COM456879056 Combi-Blocks, Inc.

Ethyl 3-amino-4-(benzylamino)benzoate

  • MDL number MFCD13562752
  • Cas Number : 848819-86-3
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9344
Availability details for
Quantity
  • Availability
  • Add to Cart