COM456879064 Combi-Blocks, Inc.

N-(4-Bromo-2,5-difluorophenyl)acetamide

  • MDL number MFCD11054214
  • Cas Number : 1065100-92-6
  • Molecular Weight : 250.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9688

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9688
Availability details for
Quantity
  • Availability
  • Add to Cart