COM456879069 Combi-Blocks, Inc.

1-BOC-3-Methylindole-2-boronic acid pinacol ester

  • MDL number MFCD18383202
  • Cas Number : 869852-13-1
  • Molecular Weight : 357.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8890

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8890
Availability details for
Quantity
  • Availability
  • Add to Cart