COM456879072 Combi-Blocks, Inc.

1-Methanesulfonyl-4-methyl-2-nitrobenzene

  • MDL number MFCD28962793
  • Cas Number : 341029-57-0
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9818
Availability details for
Quantity
  • Availability
  • Add to Cart