COM456879073 Combi-Blocks, Inc.

Methyl 3-(pyrazol-1-yl)benzoate

  • MDL number MFCD20441358
  • Cas Number : 168618-35-7
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9822
Availability details for
Quantity
  • Availability
  • Add to Cart