COM456879082 Combi-Blocks, Inc.

4-Bromo-1-(2-methylphenyl)pyrazole

  • MDL number MFCD21100064
  • Cas Number : 1337607-03-0
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9867
Availability details for
Quantity
  • Availability
  • Add to Cart