COM456879083 Combi-Blocks, Inc.

2-Chloro-4-methylsulfonyl-1-nitrobenzene

  • MDL number MFCD28962794
  • Cas Number : 60854-81-1
  • Molecular Weight : 235.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9896
Availability details for
Quantity
  • Availability
  • Add to Cart