COM456879084 Combi-Blocks, Inc.

Propanedithioamide

  • MDL number MFCD00955547
  • Cas Number : 6944-34-9
  • Molecular Weight : 134.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9905
Availability details for
Quantity
  • Availability
  • Add to Cart