COM456879085 Combi-Blocks, Inc.

5-Amino-2-bromobenzamide

  • MDL number MFCD11505332
  • Cas Number : 220583-56-2
  • Molecular Weight : 215.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9906
Availability details for
Quantity
  • Availability
  • Add to Cart