COM456879092 Combi-Blocks, Inc.

2-(3-Methoxycarbonylphenyl)benzoic acid

  • MDL number MFCD06203602
  • Cas Number : 171905-91-2
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-1231
Availability details for
Quantity
  • Availability
  • Add to Cart