COM456879093 Combi-Blocks, Inc.

6-(3,4-Methylenedioxyphenyl)nicotinic acid

  • MDL number MFCD06410426
  • Cas Number : 887976-73-0
  • Molecular Weight : 243.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6928

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YA-6928
Availability details for
Quantity
  • Availability
  • Add to Cart