COM466003078 Combi-Blocks, Inc.

4-Methylpiperidin-4-amine dihydrochloride

  • MDL number MFCD15474937
  • Cas Number : 483366-98-9
  • Molecular Weight : 187.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2453
Availability details for
Quantity
  • Availability
  • Add to Cart