COM466003094 Combi-Blocks, Inc.

2,4-dibromo-6-chloro-3-methylaniline

  • MDL number MFCD20233408
  • Cas Number : 1195139-59-3
  • Molecular Weight : 299.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6086
Availability details for
Quantity
  • Availability
  • Add to Cart