COM466003149 Combi-Blocks, Inc.

4-(imidazol-1-ylmethyl)phenylboronic acid

  • MDL number MFCD14687238
  • Cas Number : 1228183-01-4
  • Molecular Weight : 202.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4602

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4602
Availability details for
Quantity
  • Availability
  • Add to Cart