COM466003230 Combi-Blocks, Inc.

3-(n-Pentyloxy)benzoic acid

  • MDL number MFCD06801156
  • Cas Number : 110698-30-1
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1241

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1241
Availability details for
Quantity
  • Availability
  • Add to Cart