COM466003875 Combi-Blocks, Inc.

Methyl 7-bromonaphthalene-1-carboxylate

  • MDL number MFCD18413036
  • Cas Number : 93353-67-4
  • Molecular Weight : 265.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0413
Availability details for
Quantity
  • Availability
  • Add to Cart