COM466003962 Combi-Blocks, Inc.

1-Boc-2-methylazetidine-2-methanol

  • MDL number MFCD20257490
  • Cas Number : 850789-22-9
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0580
Availability details for
Quantity
  • Availability
  • Add to Cart