COM466004150 Combi-Blocks, Inc.

5-Phenyl-o-toluidine

  • MDL number MFCD00270126
  • Cas Number : 80938-67-6
  • Molecular Weight : 183.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0978
Availability details for
Quantity
  • Availability
  • Add to Cart