COM466004198 Combi-Blocks, Inc.

2,3-Dichloro-5-(methylthio)pyrazine

  • MDL number MFCD22682762
  • Cas Number : 1174517-48-6
  • Molecular Weight : 195.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1074
Availability details for
Quantity
  • Availability
  • Add to Cart