COM466004518 Combi-Blocks, Inc.

4-Chloro-2-methoxy-5-methylaniline

  • MDL number MFCD00007847
  • Cas Number : 6376-14-3
  • Molecular Weight : 171.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1756
Availability details for
Quantity
  • Availability
  • Add to Cart