COM466004580 Combi-Blocks, Inc.

2,3,5,6-Tetramethylbenzoic acid

  • MDL number MFCD00013981
  • Cas Number : 2604-45-7
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1879
Availability details for
Quantity
  • Availability
  • Add to Cart