COM466004681 Combi-Blocks, Inc.

(3R,4S)-Rel-3-hydroxy-4-piperidinemethanol

  • MDL number MFCD09910320
  • Cas Number : 21492-03-5
  • Molecular Weight : 131.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-2090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-2090
Availability details for
Quantity
  • Availability
  • Add to Cart