COM466004715 Combi-Blocks, Inc.

Methyl 3-methylenecyclobutanecarboxylate

  • MDL number MFCD01087180
  • Cas Number : 15963-40-3
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-2186

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-2186
Availability details for
Quantity
  • Availability
  • Add to Cart