COM466005203 Combi-Blocks, Inc.

Methyl 3-iodo-1-methyl-1H-indazole-6-carboxylate

  • MDL number MFCD15071434
  • Cas Number : 1041205-25-7
  • Molecular Weight : 316.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-3262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-3262
Availability details for
Quantity
  • Availability
  • Add to Cart