COM466005861 Combi-Blocks, Inc.

1,5-Dinitronaphthalene

  • MDL number MFCD00003916
  • Cas Number : 605-71-0
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-4766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-4766
Availability details for
Quantity
  • Availability
  • Add to Cart