COM466006065 Combi-Blocks, Inc.

tert-Butyl 3-bromo-6-methyl-1H-indole-1-carboxylate

  • MDL number MFCD05864782
  • Cas Number : 914349-34-1
  • Molecular Weight : 310.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5267
Availability details for
Quantity
  • Availability
  • Add to Cart