COM466006548 Combi-Blocks, Inc.

4-(Azetidin-3-yl)morpholine

  • MDL number MFCD09264459
  • Cas Number : 302355-79-9
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6267
Availability details for
Quantity
  • Availability
  • Add to Cart