COM466006575 Combi-Blocks, Inc.

(S)-6-Methoxy-2,3-dihydro-1h-inden-1-amine

  • MDL number MFCD06762392
  • Cas Number : 168903-23-9
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6322

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6322
Availability details for
Quantity
  • Availability
  • Add to Cart