COM466007052 Combi-Blocks, Inc.

(R)-Hexahydropyrrolo[1,2-a]pyrazine-1,4-dione

  • MDL number MFCD14636653
  • Cas Number : 96193-26-9
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7447
Availability details for
Quantity
  • Availability
  • Add to Cart