COM466007096 Combi-Blocks, Inc.

tert-Butyl 2,6-diazaspiro[4.5]decane-6-carboxylate hemioxalate

  • MDL number MFCD28166363
  • Cas Number : 1630907-08-2
  • Molecular Weight : 570.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7547
Availability details for
Quantity
  • Availability
  • Add to Cart