COM466007244 Combi-Blocks, Inc.

6-Boc-3-chloro-5,6,7,8-tetrahydropyrido[4,3-c]pyridazine

  • MDL number MFCD07779186
  • Cas Number : 1251015-71-0
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7865

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7865
Availability details for
Quantity
  • Availability
  • Add to Cart