COM466007738 Combi-Blocks, Inc.

1-Cyclopropylpropan-1-one

  • MDL number MFCD11934281
  • Cas Number : 6704-19-4
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-8978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-8978
Availability details for
Quantity
  • Availability
  • Add to Cart