COM466007850 Combi-Blocks, Inc.

1,2-Ethanedisulfonic acid

  • MDL number MFCD00069901
  • Cas Number : 110-04-3
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9215

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9215
Availability details for
Quantity
  • Availability
  • Add to Cart