COM466007899 Combi-Blocks, Inc.

D-Glucuronic acid sodium salt monohydrate

  • MDL number MFCD00151051
  • Cas Number : 207300-70-7
  • Molecular Weight : 234.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9325
Availability details for
Quantity
  • Availability
  • Add to Cart