COM466007963 Combi-Blocks, Inc.

5-Chlorothiazole-2-ethanone

  • MDL number MFCD09842149
  • Cas Number : 774230-95-4
  • Molecular Weight : 161.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9463
Availability details for
Quantity
  • Availability
  • Add to Cart