COM466007972 Combi-Blocks, Inc.

(2-Methoxypyrimidin-4-yl)methanamine

  • MDL number MFCD10697040
  • Cas Number : 944901-04-6
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9481
Availability details for
Quantity
  • Availability
  • Add to Cart