COM490020397 Combi-Blocks, Inc.

1'-(tert-Butoxycarbonyl)-2-oxospiro[indoline-3,4'-piperidine]-5-ylboronic acid

  • MDL number MFCD12198584
  • Cas Number : 1246372-96-2
  • Molecular Weight : 346.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3938
Availability details for
Quantity
  • Availability
  • Add to Cart