COM490020592 Combi-Blocks, Inc.

N-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-3-carboxamide

  • MDL number MFCD29043344
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1217
Availability details for
Quantity
  • Availability
  • Add to Cart