COM490020778 Combi-Blocks, Inc.

2,3,4-Trihydroxydiphenylmethane

  • MDL number MFCD08276435
  • Cas Number : 17345-66-3
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6697

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-6697
Availability details for
Quantity
  • Availability
  • Add to Cart