COM490020947 Combi-Blocks, Inc.

2-Butene-1,4-bis(triphenylphosphonium chloride)

  • MDL number MFCD00031713
  • Cas Number : 106423-29-4
  • Molecular Weight : 649.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4367

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4367
Availability details for
Quantity
  • Availability
  • Add to Cart