COM490020982 Combi-Blocks, Inc.

2-Octyne

  • MDL number MFCD00027302
  • Cas Number : 2809-67-8
  • Molecular Weight : 110.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6272
Availability details for
Quantity
  • Availability
  • Add to Cart