COM490021204 Combi-Blocks, Inc.

1-(3-Carboxyphenyl methyl) piperazine

  • MDL number MFCD06208270
  • Cas Number : 773109-07-2
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6809
Availability details for
Quantity
  • Availability
  • Add to Cart