COM490021288 Combi-Blocks, Inc.

6-Methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD00023887
  • Cas Number : 91-61-2
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1099
Availability details for
Quantity
  • Availability
  • Add to Cart