COM490021301 Combi-Blocks, Inc.

3-n-Butyl-2,4-pentanedione

  • MDL number MFCD00026235
  • Cas Number : 1540-36-9
  • Molecular Weight : 156.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2927
Availability details for
Quantity
  • Availability
  • Add to Cart