COM490021394 Combi-Blocks, Inc.

3-Ethyl-2-pentene

  • MDL number MFCD00027079
  • Cas Number : 816-79-5
  • Molecular Weight : 98.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2749

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2749
Availability details for
Quantity
  • Availability
  • Add to Cart