COM490021417 Combi-Blocks, Inc.

Cis-13-docosenol

  • MDL number MFCD00055936
  • Cas Number : 629-98-1
  • Molecular Weight : 324.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4254
Availability details for
Quantity
  • Availability
  • Add to Cart