COM490021419 Combi-Blocks, Inc.

5-(Benzoylamino)valeric acid

  • MDL number MFCD00021790
  • Cas Number : 15647-47-9
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4391
Availability details for
Quantity
  • Availability
  • Add to Cart