COM490021434 Combi-Blocks, Inc.

5-Norbornene-2-carbonitrile

  • MDL number MFCD00167563
  • Cas Number : 95-11-4
  • Molecular Weight : 119.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5758
Availability details for
Quantity
  • Availability
  • Add to Cart